missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 4-(Dimethylamino)benzoate 98.0+%, TCI America™

Catalog No. p-7126982
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187125ML 25 mL
D1871100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D187125ML Supplier TCI America Supplier No. D187125ML

Chemical Identifiers

CAS 21245-02-3
Molecular Formula C17H27NO2
Molecular Weight (g/mol) 277.408
MDL Number MFCD00017526
InChI Key WYWZRNAHINYAEF-UHFFFAOYSA-N
Synonym 2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown
PubChem CID 30541
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Color Yellow
Boiling Point 325°C
Quantity 25 mL
Formula Weight 277.41
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 4-(Dimethylamino)benzoate
EINECSNumber (3)-1436
TSCA Yes