missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexanoyl Chloride 98.0+%, TCI America™
$55.92 - $287.06
Chemical Identifiers
| CAS | 760-67-8 |
|---|---|
| Molecular Formula | C8H15ClO |
| Molecular Weight (g/mol) | 162.657 |
| MDL Number | MFCD00000718 |
| InChI Key | WFSGQBNCVASPMW-UHFFFAOYSA-N |
| Synonym | hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride |
| PubChem CID | 79103 |
| IUPAC Name | 2-ethylhexanoyl chloride |
| SMILES | CCCCC(CC)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E033625ML
|
TCI America
E033625ML |
25 mL |
Each for $55.92
|
|
|||||
|
E0336500ML
|
TCI America
E0336500ML |
500 mL |
Each for $287.06
|
|
|||||
Chemical Identifiers
| 760-67-8 | |
| 162.657 | |
| WFSGQBNCVASPMW-UHFFFAOYSA-N | |
| 79103 | |
| CCCCC(CC)C(=O)Cl |
| C8H15ClO | |
| MFCD00000718 | |
| hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride | |
| 2-ethylhexanoyl chloride |
Specifications
| 760-67-8 | |
| 101°C | |
| MFCD00000718 | |
| 3265 | |
| WFSGQBNCVASPMW-UHFFFAOYSA-N | |
| 2-ethylhexanoyl chloride | |
| 79103 | |
| ≥98.0% (GC,T) | |
| 2-Ethylhexanoyl Chloride |
| Yellow | |
| C8H15ClO | |
| 25 mL | |
| hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride | |
| CCCCC(CC)C(=O)Cl | |
| 162.657 | |
| 162.66 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0632
RTECSNumber : MP4525000
TSCA : Yes