missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoyl Chloride 98.0+%, TCI America™

Catalog No. E033625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E033625ML 25 mL
E0336500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E033625ML Supplier TCI America Supplier No. E033625ML
Only null left

Chemical Identifiers

CAS 760-67-8
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.657
MDL Number MFCD00000718
InChI Key WFSGQBNCVASPMW-UHFFFAOYSA-N
Synonym hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride
PubChem CID 79103
IUPAC Name 2-ethylhexanoyl chloride
SMILES CCCCC(CC)C(=O)Cl

Specifications

Color Yellow
Boiling Point 101°C
Quantity 25 mL
UN Number 3265
Formula Weight 162.66
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanoyl Chloride
EINECSNumber (2)-0632
RTECSNumber MP4525000
TSCA Yes