Learn More
2-Ethylhexane-1,3-diol, 97%
CAS: 94-96-2 | C8H18O2 | 146.23 g/mol
Supplier: Thermo Scientific Chemicals A108980D
Description
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.
Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
Specifications
| 2-Ethylhexane-1,3-diol | |
| -40°C | |
| Odorless | |
| CH3CH2CH2CHOHCH(CH2CH3)CH2OH | |
| 10,000 mL | |
| Soluble in water (42g/L at 20°C). Soluble in alcohol and ether. | |
| CCCC(O)C(CC)CO | |
| 146.23 | |
| CHEBI:34273 | |
| 97% |
| 94-96-2 | |
| 241°C to 249°C | |
| C8H18O2 | |
| MFCD00004578 | |
| 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8 | |
| RWLALWYNXFYRGW-UHFFFAOYNA-N | |
| 2-ethylhexane-1,3-diol | |
| 7211 | |
| 146.23 |
Chemical Identifiers
| 94-96-2 | |
| 146.23 | |
| RWLALWYNXFYRGW-UHFFFAOYNA-N | |
| 7211 | |
| 2-ethylhexane-1,3-diol |
| C8H18O2 | |
| MFCD00004578 | |
| 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8 | |
| CHEBI:34273 | |
| CCCC(O)C(CC)CO |
Safety and Handling
GHS H Statement
H318-H302-H312
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P333+P313-P363-P501c
H302-H312-H317-H318
EINECSNumber : 202-377-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only