missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal 95.0+%, TCI America™

Catalog No. E012525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E012525ML 25 mL
E0125500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E012525ML Supplier TCI America Supplier No. E012525ML

Chemical Identifiers

CAS 123-05-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00006987
InChI Key LGYNIFWIKSEESD-UHFFFAOYSA-N
Synonym hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
PubChem CID 31241
IUPAC Name 2-ethylhexanal
SMILES CCCCC(CC)C=O

Specifications

Melting Point -85°C
Color Colorless
Boiling Point 163°C
Quantity 25 mL
UN Number 1191
Formula Weight 128.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanal
EINECSNumber (2)-0494
RTECSNumber MN7525000
TSCA Yes