missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal, 96%

Catalog No. p-129956
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410092500 250 g Glass bottle
AC410090010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC410092500 Supplier Thermo Scientific Chemicals Supplier No. 410092500
Only null left

CAS: 123-05-7 | C8H16O | 128.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-05-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.21
MDL Number MFCD00006987
InChI Key LGYNIFWIKSEESD-UHFFFAOYSA-N
Synonym hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
PubChem CID 31241
IUPAC Name 2-ethylhexanal
SMILES CCCCC(CC)C=O

Specifications

Acidity 8mg KOH/g max.
CAS Min % 95.0
CAS Max % 100.0
Melting Point -90°C
Color Colorless
Density 0.8195g/mL
Boiling Point 162.0°C to 165.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4145 to 1.4175
Linear Formula CH3(CH2)3CH(C2H5)CHO
Quantity 250 g
Specific Gravity 0.8195
Solubility Information Solubility in water: < 0.2g/L (20°C). Other solubilities: misccible with most organic solvents
Formula Weight 128.21
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexanal, pract., 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Obtain special instructions before use.
Wea
EINECSNumber 204-596-5
RTECSNumber MN7525000
TSCA TSCA

RUO – Research Use Only