missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylfuran 98.0+%, TCI America™

Catalog No. E045410ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E045410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E045410ML Supplier TCI America Supplier No. E045410ML

Chemical Identifiers

CAS 3208-16-0
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00003259
InChI Key HLPIHRDZBHXTFJ-UHFFFAOYSA-N
Synonym furan, 2-ethyl, alpha-ethylfuran, 2-ethyloxole, 2-ethyl furan, ccris 4387, unii-t8o6j71t9o, fema no. 3673, furan, .alpha.-ethyl, a-ethylfuran, 2-ethyl furane
PubChem CID 18554
IUPAC Name 2-ethylfuran
SMILES CCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 10 mL
UN Number 1993
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylfuran