missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%

Catalog No. p-7035922
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54151MD 250 mg
AAH5415103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH54151MD Supplier Thermo Scientific Chemicals Supplier No. H54151MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1006876-28-3 | C14H21BN2O3 | 276.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1006876-28-3
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.143
MDL Number MFCD13191367
InChI Key TVMRHMZREIYXSR-UHFFFAOYSA-N
Synonym n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 6-ethylcarbamoyl pyridine-3-boronic acid pinacol ester, 2-ethylcarbamoyl pyridine-5-boronic acid pinacol ester, 2-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester, 2-n-ethylaminocarbonyl pyridine-5-boronic acid pinacol ester, 2-pyridinecarboxamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl picolinamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine-2-carboxylic acid ethylamide, n-ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarboxamide
PubChem CID 59868917
IUPAC Name N-ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)NCC

Specifications

Melting Point 52°C to 54°C
Quantity 250 mg
Formula Weight 276.14
Percent Purity 96%
Chemical Name or Material 2-(Ethylcarbamoyl)pyridine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only