missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanthraquinone 98.0+%, TCI America™

$33.56 - $300.11

Chemical Identifiers

CAS 84-51-5
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00001237
InChI Key SJEBAWHUJDUKQK-UHFFFAOYSA-N
Synonym 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
PubChem CID 6772
IUPAC Name 2-ethylanthracene-9,10-dione
SMILES CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E006325G
View Documents
TCI America
E006325G
25 g N/A
Only null left
 
E0063500G
View Documents
TCI America
E0063500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

84-51-5
236.27
SJEBAWHUJDUKQK-UHFFFAOYSA-N
6772
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
C16H12O2
MFCD00001237
2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
2-ethylanthracene-9,10-dione

Specifications

84-51-5
190°C
MFCD00001237
2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
236.27
236.27
Crystalline Powder
108°C
C16H12O2
25 g
SJEBAWHUJDUKQK-UHFFFAOYSA-N
2-ethylanthracene-9,10-dione
6772
≥98.0% (GC)
2-Ethylanthraquinone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0687

RTECSNumber : CB0525000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'