missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanthraquinone 98.0+%, TCI America™

Catalog No. E006325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006325G 25 g
E0063500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E006325G Supplier TCI America Supplier No. E006325G

Chemical Identifiers

CAS 84-51-5
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00001237
InChI Key SJEBAWHUJDUKQK-UHFFFAOYSA-N
Synonym 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
PubChem CID 6772
IUPAC Name 2-ethylanthracene-9,10-dione
SMILES CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 108°C
Boiling Point 190°C
Quantity 25 g
Formula Weight 236.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethylanthraquinone
EINECSNumber (4)-0687
RTECSNumber CB0525000
TSCA Yes