missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylanthraquinone, 98%

Catalog No. AAL0720606
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0720606 5 g
AAL0720622 100 g
AAL0720636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0720606 Supplier Thermo Scientific Chemicals Supplier No. L0720606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84-51-5 | C16H12O2 | 236.27 g/mol

2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 84-51-5
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00001237
InChI Key SJEBAWHUJDUKQK-UHFFFAOYSA-N
Synonym 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone
PubChem CID 6772
IUPAC Name 2-ethylanthracene-9,10-dione
SMILES CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 107°C to 111°C
Boiling Point 180°C to 190°C
Flash Point >210°C (410°F)
Quantity 5 g
Beilstein 1969873
Solubility Information Insoluble in water.
Formula Weight 236.27
Percent Purity 98%
Chemical Name or Material 2-Ethylanthraquinone
EINECSNumber 201-535-4
RTECSNumber CB0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only