missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-5-nitroaniline 98.0+%, TCI America™

Catalog No. E10851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10851G 1 g
E10855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10851G Supplier TCI America Supplier No. E10851G
Only null left

Chemical Identifiers

CAS 20191-74-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00034056
InChI Key MMZWMCKTKJKIMC-UHFFFAOYSA-N
PubChem CID 4155478
IUPAC Name 2-ethyl-5-nitroaniline
SMILES CCC1=C(C=C(C=C1)[N+](=O)[O-])N

Specifications

Melting Point 60°C
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-5-nitroaniline