missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, 99%

Catalog No. p-7031201
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3218706 5 g
AAH3218714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3218706 Supplier Thermo Scientific Chemicals Supplier No. H3218706
Only null left

CAS: 4359-46-0 | C6H12O2 | 116.16 g/mol

2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1,3-dioxolane, cis + trans, is used as a flavouring agent in food industry.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 4359-46-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD16622267
InChI Key CSZCLQLJVFLXLI-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-ethyl-4-methyl, 1,3-dioxolane,2-ethyl-4-methyl, cis, 1,2-propanediol acetal, 1,3-dioxolane, 2-ethyl-4-methyl-, cis, cis-2-thyl-4-methyl-1,3-dioxolane, 2-ethyl-4-methyl-1,3-dioxolane, cis + trans, solution in acetonitrile 1000mg/l
PubChem CID 92210
IUPAC Name 2-ethyl-4-methyl-1,3-dioxolane
SMILES CCC1OCC(O1)C

Specifications

Density 0.904
Boiling Point 117°C
Flash Point 29°C (84°F)
Odor Characteristic
Refractive Index 1.403
Quantity 5 g
UN Number UN1993
Solubility Information Slightly soluble in water. Soluble in alcohol.
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 2-Ethyl-4-methyl-1,3-dioxolane, Cis + Trans
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only