missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone 97.0+%, TCI America™

Catalog No. E042810G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E042810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E042810G Supplier TCI America Supplier No. E042810G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27538-10-9
Molecular Formula C7H10O3
Molecular Weight (g/mol) 142.15
MDL Number MFCD00191360
InChI Key GWCRPYGYVRXVLI-UHFFFAOYNA-N
PubChem CID 33931
IUPAC Name 2-ethyl-4-hydroxy-5-methyl-2,3-dihydrofuran-3-one
SMILES CCC1OC(C)=C(O)C1=O

Specifications

Color Yellow
Quantity 10 g
Formula Weight 142.15
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-4-hydroxy-5-methyl-3(2H)-furanone
RTECSNumber LU4250000
TSCA Yes