missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. p-7129341
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11161G 1 g
E11165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11161G Supplier TCI America Supplier No. E11161G

Chemical Identifiers

CAS 82954-89-0
Molecular Formula C8H17BO2
Molecular Weight (g/mol) 156.032
MDL Number MFCD19441153
InChI Key NPUBDPDASOEIOA-UHFFFAOYSA-N
Synonym (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethane, Ethylboronic Acid Pinacol Ester
PubChem CID 10942698
IUPAC Name 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC

Specifications

Color Colorless
Quantity 1 g
UN Number 3272
Formula Weight 156.03
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator