missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-3-methylpyrazine 98.0+%, TCI America™
Supplier: TCI America E036125ML
Specifications
| 2-Ethyl-3-methylpyrazine | |
| Yellow | |
| C7H10N2 | |
| 25 mL | |
| pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci | |
| CCC1=NC=CN=C1C | |
| 122.17 | |
| 122.17 | |
| Liquid |
| 15707-23-0 | |
| 70°C | |
| MFCD00006150 | |
| 1993 | |
| LNIMMWYNSBZESE-UHFFFAOYSA-N | |
| 2-ethyl-3-methylpyrazine | |
| 27457 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 15707-23-0 | |
| 122.17 | |
| LNIMMWYNSBZESE-UHFFFAOYSA-N | |
| 27457 | |
| CCC1=NC=CN=C1C |
| C7H10N2 | |
| MFCD00006150 | |
| pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci | |
| 2-ethyl-3-methylpyrazine |
Safety and Handling
RTECSNumber : UQ3335000
TSCA : Yes