Learn More
2-Ethyl-3-methylpyrazine, 98+%
CAS: 15707-23-0 | C7H10N2 | 122.17 g/mol
$65.32 - $605.49
Chemical Identifiers
| CAS | 15707-23-0 |
|---|---|
| Molecular Formula | C7H10N2 |
| Molecular Weight (g/mol) | 122.17 |
| MDL Number | MFCD00006150 |
| InChI Key | LNIMMWYNSBZESE-UHFFFAOYSA-N |
| Synonym | pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci |
| PubChem CID | 27457 |
| IUPAC Name | 2-ethyl-3-methylpyrazine |
| SMILES | CCC1=NC=CN=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1109206
|
Thermo Scientific Chemicals
A1109206 |
5 g |
N/A
|
|
|||||
|
AAA1109214
|
Thermo Scientific Chemicals
A1109214 |
25 g |
N/A
|
|
|||||
|
AAA1109222
|
Thermo Scientific Chemicals
A1109222 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 15707-23-0 | |
| 122.17 | |
| LNIMMWYNSBZESE-UHFFFAOYSA-N | |
| 27457 | |
| CCC1=NC=CN=C1C |
| C7H10N2 | |
| MFCD00006150 | |
| pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci | |
| 2-ethyl-3-methylpyrazine |
Specifications
| 15707-23-0 | |
| 169°C to 171°C | |
| Characteristic | |
| 1.503 | |
| 5 g | |
| 956775 | |
| LNIMMWYNSBZESE-UHFFFAOYSA-N | |
| 2-ethyl-3-methylpyrazine | |
| 27457 | |
| ≥98% |
| 0.987 | |
| 57°C (134°F) | |
| C7H10N2 | |
| MFCD00006150 | |
| UN1993 | |
| pyrazine, 2-ethyl-3-methyl, 3-ethyl-2-methylpyrazine, 2-methyl-3-ethylpyrazine, 2-ethyl-3-methyl pyrazine, pyrazine, ethylmethyl, 2-ethyl-3-methyl-pyrazine, unii-9gf35mk66u, fema no. 3155, pyrazine, 3-ethyl-2-methyl, 2-ethyl-3-methylpyrazine, 9ci | |
| CCC1=NC=CN=C1C | |
| 122.17 | |
| 122.17 | |
| 2-Ethyl-3-methylpyrazine |
Safety and Handling
GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 239-799-8
RTECSNumber : UQ3335000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only