missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-oxazoline 98.0+%, TCI America™

Catalog No. E0373500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0373500G 500 g
E037325G 25 g
E0373100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0373500G Supplier TCI America Supplier No. E0373500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10431-98-8
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00005302
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC1=NCCO1

Specifications

Color Colorless
Boiling Point 128°C
Quantity 500 g
UN Number 1993
Formula Weight 99.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-2-oxazoline
EINECSNumber (5)-5627
TSCA Yes