missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-oxazoline, 99+%, Thermo Scientific™

Catalog No. AC159980025
Change view
Click to view available options
2.5Kg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC159980025 2.5Kg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159980025 Supplier Thermo Scientific Chemicals Supplier No. 159980025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 10431-98-8
Density 0.9800g/mL
Boiling Point 128.4°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C5H9NO
Refractive Index 1.4360 to 1.4380
MDL Number MFCD00005302
Quantity 2.5kg
Specific Gravity 0.98
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
Solubility Information Solubility in water: miscible
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES CCC1=NCCO1
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol) 99.13
PubChem CID 66412
Viscosity 0.88 mPa.s (20°C)
Formula Weight 99.13
Percent Purity 99+%
Chemical Name or Material 2-Ethyl-2-oxazoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 233-912-4

RUO – Research Use Only