missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-oxazoline, 99+%

Catalog No. AC159981000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159981000 100 g Glass bottle
AC159985000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159981000 Supplier Thermo Scientific Chemicals Supplier No. 159981000

CAS: 10431-98-8 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10431-98-8
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00005302
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC1=NCCO1

Specifications

Melting Point -62.0°C
Density 0.9800g/mL
Boiling Point 128.4°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4360 to 1.4380
Quantity 100 g
Specific Gravity 0.98
Solubility Information Solubility in water: miscible
Viscosity 0.88 mPa.s (20°C)
Formula Weight 99.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-2-oxazoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 233-912-4

RUO – Research Use Only