missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2-Ethyl-2-oxazoline, 99%

Catalog No. AAB2328736
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2328736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2328736 Supplier Thermo Scientific™ Supplier No. B2328736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10431-98-8 | C5H9NO | 99.13 g/mol

2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 10431-98-8
Melting Point -62°
Density 0.981
Boiling Point 128-129°
Flash Point 29°(84°F)
Molecular Formula C5H9NO
Quantity 500 g
MDL Number MFCD00005302
UN Number UN2734
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES CCC1=NCCO1
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol) 99.13
Formula Weight 99.13
Percent Purity 99%
Physical Form Liquid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 233-912-4