missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-methyl-1,3-dioxolane 98.0+%, TCI America™

Catalog No. E051525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E051525ML 25 mL
E0515500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E051525ML Supplier TCI America Supplier No. E051525ML

Chemical Identifiers

CAS 126-39-6
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00014108
InChI Key UPZFLZYXYGBAPL-UHFFFAOYSA-N
Synonym 2-methyl-2-ethyl-1,3-dioxolane, ethyleneacetic acid, 2-butanone, cyclic ethylene acetal, 2-ethyl-2-methyldioxolane, 2-methyl-2-ethyldioxolane, 1,3-dioxolane, ethylmethyl, 1,3-dioxolane, 2-ethyl-2-methyl, unii-y488vjj1fv, 2-butanone, cyclic 1,2-ethanediyl acetal, y488vjj1fv
PubChem CID 61067
IUPAC Name 2-ethyl-2-methyl-1,3-dioxolane
SMILES CCC1(OCCO1)C

Specifications

Color Yellow
Boiling Point 117°C
Quantity 25 mL
UN Number 1993
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-2-methyl-1,3-dioxolane
RTECSNumber JH7700000
TSCA Yes