missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

Catalog No. aaa130840e
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 g
2500 g
500 g
Catalog No. Quantity
AAA130840E 2500 g
AAA1308436 500 g
AAA130840C 10,000 g
3 options

Catalog No. AAA130840E

Supplier: Thermo Scientific Chemicals A130840E

Only null left

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 77-99-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00004694
InChI Key ZJCCRDAZUWHFQH-UHFFFAOYSA-N
Synonym trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
PubChem CID 6510
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
SMILES CCC(CO)(CO)CO

Specifications

Melting Point 59°C to 60°C
Boiling Point 296°C to 298°C
Flash Point 172°C (341°F)
Quantity 2500 g
Beilstein 1698309
Solubility Information Soluble in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol
EINECSNumber 201-074-9
RTECSNumber TY6470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only