Learn More
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
Description
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol acts as a precursor to alkyd resins, high-gloss coatings and ion exchange resins. It is also employed as a multifunctional monomer utilized for the production of coatings, ethoxylated and propoxylated trimethylolpropane derivatives. Further, it serves as a building block in the polymer industry.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 59°C to 60°C |
| Boiling Point | 296°C to 298°C |
| Flash Point | 172°C (341°F) |
| Quantity | 10,000 g |
| Beilstein | 1698309 |
| Solubility Information | Soluble in water. |
| Formula Weight | 134.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Ethyl-2-(hydroxymethyl)-1,3-propanediol |
Safety and Handling
| EINECSNumber | 201-074-9 |
| RTECSNumber | TY6470000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only