missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-hexene 95.0+%, TCI America™

Catalog No. p-7128851
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E012410ML 10 mL
E012425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E012410ML Supplier TCI America Supplier No. E012410ML

Chemical Identifiers

CAS 1632-16-2
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00027242
InChI Key XTVRLCUJHGUXCP-UHFFFAOYSA-N
PubChem CID 15404
IUPAC Name 3-methylideneheptane
SMILES CCCCC(=C)CC

Specifications

Color Colorless
Boiling Point 120°C
Quantity 10 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-hexene
EINECSNumber (2)-0024
RTECSNumber MP6825000
TSCA Yes