missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-hexanethiol 98.0+%, TCI America™

Catalog No. p-7129291
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E103125G 25 g
E1031100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E103125G Supplier TCI America Supplier No. E103125G

Chemical Identifiers

CAS 7341-17-5
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.292
MDL Number MFCD00039654
InChI Key UCJMHYXRQZYNNL-UHFFFAOYSA-N
Synonym 2-Ethylhexyl Mercaptan
PubChem CID 110968
IUPAC Name 2-ethylhexane-1-thiol
SMILES CCCCC(CC)CS

Specifications

Melting Point -40°C
Color Colorless
Boiling Point 67°C
Quantity 25 g
UN Number 3336
Formula Weight 146.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-hexanethiol
EINECSNumber (2)-0464
TSCA Yes