missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-butene 97.0+%, TCI America™

Catalog No. E006725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006725ML 25 mL
E0067100ML 100 mL
E0067500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E006725ML Supplier TCI America Supplier No. E006725ML

Chemical Identifiers

CAS 760-21-4
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009341
InChI Key RYKZRKKEYSRDNF-UHFFFAOYSA-N
Synonym 2-ethyl-1-butene, 3-methylenepentane, pentane, 3-methylene, 2-ethylbut-1-ene, 1-butene, 2-ethyl, unii-o5mh5v756e, 1,1-diethylethene, acmc-1blm9, 1-butene, 2-ethyl-8ci
PubChem CID 12970
IUPAC Name 3-methylidenepentane
SMILES CCC(=C)CC

Specifications

Color Yellow
Boiling Point 64°C
Quantity 25 mL
UN Number 3295
Formula Weight 84.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-butene