missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1,3-hexanediol, 99%, mixture of isomers

Catalog No. p-129901
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118512500 250 mL Glass bottle
AC118510010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC118512500 Supplier Thermo Scientific Chemicals Supplier No. 118512500
Only null left

CAS: 94-96-2 | C8H18O2 | 146.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-96-2
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004578
InChI Key RWLALWYNXFYRGW-UHFFFAOYNA-N
Synonym 2-ethyl-1,3-hexanediol, ethohexadiol, 1,3-hexanediol, 2-ethyl, octylene glycol, ethyl hexanediol, carbide 6-12, repellent 612, rutgers 612, 6-12-insect repellent, diol-kyowa 8
PubChem CID 7211
ChEBI CHEBI:34273
IUPAC Name 2-ethylhexane-1,3-diol
SMILES CCCC(O)C(CC)CO

Specifications

Acidity 0.05mg KOH/g max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -40°C
Density 0.9422g/mL
Boiling Point 243°C
Flash Point 136°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4495 to 1.4525
Linear Formula CH3CH2CH2CH(OH)CH(C2H5)CH2OH
Quantity 250 mL
Specific Gravity 0.9422
Merck Index 15, 3797
Solubility Information Solubility in water: 42g/L (20°C). Other solubilities: soluble in ethanol,isopropanol,propylene glycol,,castor oil and ether
Viscosity 323 mPa.s (20°C)
Formula Weight 146.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1, 3-hexanediol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 202-377-9
RTECSNumber MO2625000
TSCA TSCA

RUO – Research Use Only