missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-1,3-cyclopentanedione 97.0+%, TCI America™
$44.28 - $44.28
Chemical Identifiers
| CAS | 823-36-9 |
|---|---|
| Molecular Formula | C7H10O2 |
| Molecular Weight (g/mol) | 126.155 |
| MDL Number | MFCD00044191 |
| InChI Key | YDFBIBUYOUFJMR-UHFFFAOYSA-N |
| Synonym | 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione |
| PubChem CID | 69978 |
| IUPAC Name | 2-ethylcyclopentane-1,3-dione |
| SMILES | CCC1C(=O)CCC1=O |
Chemical Identifiers
| 823-36-9 | |
| 126.155 | |
| YDFBIBUYOUFJMR-UHFFFAOYSA-N | |
| 69978 | |
| CCC1C(=O)CCC1=O |
| C7H10O2 | |
| MFCD00044191 | |
| 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione | |
| 2-ethylcyclopentane-1,3-dione |
Specifications
| 823-36-9 | |
| White-Yellow | |
| MFCD00044191 | |
| 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione | |
| CCC1C(=O)CCC1=O | |
| 126.155 | |
| 126.16 | |
| Crystalline Powder |
| 175°C | |
| C7H10O2 | |
| 5 g | |
| YDFBIBUYOUFJMR-UHFFFAOYSA-N | |
| 2-ethylcyclopentane-1,3-dione | |
| 69978 | |
| ≥97.0% (GC,T) | |
| 2-Ethyl-1,3-cyclopentanedione |
Safety and Handling
TSCA : No