missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1,3-cyclopentanedione, 98%

Catalog No. AAB2025414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2025414 25 g
AAB2025406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2025414 Supplier Thermo Scientific Chemicals Supplier No. B2025414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 823-36-9 | C7H10O2 | 126.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 823-36-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00044191
InChI Key YDFBIBUYOUFJMR-UHFFFAOYSA-N
Synonym 2-ethyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-ethyl, unii-91a82te3os, acmc-209poa, 2-ethyl-cyclopentan-1,3-dione, 2-ethyl-cyclopentane-1,3-dione
PubChem CID 69978
IUPAC Name 2-ethylcyclopentane-1,3-dione
SMILES CCC1C(=O)CCC1=O

Specifications

Melting Point 168°C to 170°C
Quantity 25 g
Beilstein 1860068
Formula Weight 126.16
Percent Purity 99%
Chemical Name or Material 2-Ethyl-1,3-cyclopentanedione
EINECSNumber 212-512-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only