missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxythiazole 98.0+%, TCI America™

Catalog No. p-7129095
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06695G 5 g
E066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06695G Supplier TCI America Supplier No. E06695G
Only null left

Chemical Identifiers

CAS 15679-19-3
Molecular Formula C5H7NOS
Molecular Weight (g/mol) 129.177
InChI Key NDUWJHRKDYXRAD-UHFFFAOYSA-N
PubChem CID 61809
IUPAC Name 2-ethoxy-1,3-thiazole
SMILES CCOC1=NC=CS1

Specifications

Color Yellow
Boiling Point 160°C
Quantity 5 g
Formula Weight 129.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxythiazole