missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-3-carboxaldehyde, 97%

Catalog No. AAH5008806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5008806 5 g
AAH5008803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5008806 Supplier Thermo Scientific Chemicals Supplier No. H5008806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 885278-07-9 | C8H9NO2 | 151.17 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 885278-07-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD06804559
InChI Key WHEFRNNNELKVRQ-UHFFFAOYSA-N
Synonym 2-ethoxynicotinaldehyde, 2-ethoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-ethoxy, 2-ethoxy-3-pyridinecarboxaldehyde, acmc-20ac3a, 2-ethoxy-pyridine-3-carbaldehyde
PubChem CID 25067327
IUPAC Name 2-ethoxypyridine-3-carbaldehyde
SMILES CCOC1=C(C=O)C=CC=N1

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 2-Ethoxypyridine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only