missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-3-boronic acid, 98%

Catalog No. p-7048170
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL20422MD 250 mg
AAL2042203 1 g
AAL2042206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL20422MD Supplier Thermo Scientific Chemicals Supplier No. L20422MD

CAS: 854373-97-0 | C7H10BNO3 | 166.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 854373-97-0
Molecular Formula C7H10BNO3
Molecular Weight (g/mol) 166.97
MDL Number MFCD06201036
InChI Key FXUMKSCYKPOZOO-UHFFFAOYSA-N
Synonym 2-ethoxypyridine-3-boronic acid, 2-ethoxy-3-pyridineboronic acid, 2-ethoxypyridin-3-yl boronic acid, 2-ethoxy-3-pyridyl boronic acid, 3-borono-2-ethoxypyridine, pubchem5119, acmc-209q5z, 2-ethoxypyridine-3-boronicacid, 2-ethoxy-3-pyridinyl boronic acid, 2-ethoxy-pyridin-3-yl-boronic acid
PubChem CID 11309733
IUPAC Name (2-ethoxypyridin-3-yl)boronic acid
SMILES CCOC1=NC=CC=C1B(O)O

Specifications

Quantity 250 mg
Formula Weight 166.97
Percent Purity 98%
Chemical Name or Material 2-Ethoxypyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only