missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenol, 98%, Thermo Scientific™

Chemical Identifiers

CAS 94-71-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002187
InChI Key MOEFFSWKSMRFRQ-UHFFFAOYSA-N
Synonym o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
PubChem CID 66755
IUPAC Name 2-ethoxyphenol
SMILES CCOC1=CC=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC370030050
SDS View Documents
Thermo Scientific Chemicals
370030050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC370035000
SDS View Documents
Thermo Scientific Chemicals
370035000
500g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

94-71-3
138.17
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
66755
CCOC1=CC=CC=C1O
C8H10O2
MFCD00002187
o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
2-ethoxyphenol

Specifications

94-71-3
100.0
216.0°C to 217.0°C
Authentic
Glass bottle
1.5280 to 1.5300
MFCD00002187
1.09
MOEFFSWKSMRFRQ-UHFFFAOYSA-N
2-ethoxyphenol
66755
98%
97.5
1.0900g/mL
91°C
97.5% min. (GC)
C8H10O2
C2H5OC6H4OH
5g
o-ethoxyphenol, phenol, 2-ethoxy, guaethol, guethol, 2-ethyloxyphenol, guaiethol, pyrocatechol monoethyl ether, catechol monoethyl ether, phenol, o-ethoxy, unii-878iw8p9pw
CCOC1=CC=CC=C1O
138.17
138.17
2-Ethoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 202-358-5

SDS
Documents

Documents

RUO – Research Use Only