missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Cyanoacetate 98.0+%, TCI America™

Catalog No. p-7125152
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C175625G 25 g
C1756500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C175625G Supplier TCI America Supplier No. C175625G

Chemical Identifiers

CAS 32804-77-6
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00071546
InChI Key UMCLCZMPTREESK-UHFFFAOYSA-N
Synonym Cyanoacetic Acid 2-Ethoxyethyl Ester
PubChem CID 122967
IUPAC Name 2-ethoxyethyl 2-cyanoacetate
SMILES CCOCCOC(=O)CC#N

Specifications

Color Colorless
Quantity 25 g
Formula Weight 157.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Cyanoacetate
EINECSNumber (2)-3542
TSCA No