missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester, 96%

Catalog No. p-7034623
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52312MD 250 mg
AAH5231203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52312MD Supplier Thermo Scientific Chemicals Supplier No. H52312MD
Only null left

CAS: 1073338-92-7 | C13H19BO5 | 266.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1073338-92-7
Molecular Formula C13H19BO5
Molecular Weight (g/mol) 266.10
MDL Number MFCD11855980
InChI Key UMMKKZJPSPIDGN-UHFFFAOYSA-N
PubChem CID 46739337
IUPAC Name ethyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carboxylate
SMILES CCOC(=O)C1=CC=C(O1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Synonym ethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-ethoxycarbonyl furan-2-boronic acid, pinacol ester, 5-ethoxycarbonyl furan-2-boronic acid pinacol ester, 2-ethoxycarbonyl furan-5-boronic acid pinacol ester, ethyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, ethyl5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-furan-2-carboxylic acid ethyl ester
Formula Weight 266.1
Percent Purity 96%
Chemical Name or Material 2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only