missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
$315.99 - $315.99
Chemical Identifiers
| CAS | 269409-99-6 |
|---|---|
| Molecular Formula | C15H21BO4 |
| Molecular Weight (g/mol) | 276.139 |
| MDL Number | MFCD03453660 |
| InChI Key | RFBZWPFBCXBBJS-UHFFFAOYSA-N |
| Synonym | ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate |
| PubChem CID | 2773962 |
| IUPAC Name | ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3094106
|
Thermo Scientific Chemicals
H3094106 |
5 g |
Each for $315.99
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 269409-99-6 | |
| 276.139 | |
| RFBZWPFBCXBBJS-UHFFFAOYSA-N | |
| 2773962 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC |
| C15H21BO4 | |
| MFCD03453660 | |
| ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate | |
| ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate |
Specifications
| 269409-99-6 | |
| C15H21BO4 | |
| 5 g | |
| RFBZWPFBCXBBJS-UHFFFAOYSA-N | |
| ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate | |
| 2773962 | |
| 97% |
| 57°C to 62°C | |
| MFCD03453660 | |
| ethyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid pinacol ester, ethyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-ethoxycarbonylphenylboronic acid, pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid ethyl ester, ethyl 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-carboethoxyphenylboronic acid pinacol ester, 2-ethoxycarbonyl benzeneboronic acid pinacol ester, ethyl 2-4,4,5,5-tetramethyl 1,3,2 dioxaborolan-2-yl benzoate | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C(=O)OCC | |
| 276.139 | |
| 276.14 | |
| 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only