missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamide 98.0+%, TCI America™

Catalog No. p-7128897
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E022225G 25 g
E0222500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E022225G Supplier TCI America Supplier No. E022225G
Only null left

Chemical Identifiers

CAS 938-73-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007977
InChI Key SBNKFTQSBPKMBZ-UHFFFAOYSA-N
Synonym ethenzamide, benzamide, 2-ethoxy, o-ethoxybenzamide, ethbenzamide, etenzamide, ethenzamid, etamide, ethosalicyl, etosalicil, etosalicyl
PubChem CID 3282
IUPAC Name 2-ethoxybenzamide
SMILES CCOC1=CC=CC=C1C(N)=O

Specifications

Melting Point 131°C
Color White
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethoxybenzamide
EINECSNumber (3)-1679
RTECSNumber CV4900000
TSCA Yes