missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzamide, 97%

Catalog No. p-125930
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117870050 5 g Glass bottle
AC117871000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117870050 Supplier Thermo Scientific Chemicals Supplier No. 117870050

CAS: 938-73-8 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 938-73-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00007977
InChI Key SBNKFTQSBPKMBZ-UHFFFAOYSA-N
Synonym ethenzamide, benzamide, 2-ethoxy, o-ethoxybenzamide, ethbenzamide, etenzamide, ethenzamid, etamide, ethosalicyl, etosalicil, etosalicyl
PubChem CID 3282
IUPAC Name 2-ethoxybenzamide
SMILES CCOC1=CC=CC=C1C(N)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 129.0°C to 134.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C2H5OC6H4CONH2
Quantity 5 g
Beilstein 10, 93
Merck Index 15, 3786
Solubility Information Solubility in water: slightly soluble in water.
Formula Weight 165.19
Percent Purity 97%
Physical Form Granular Crystalline Powder
Chemical Name or Material 2-Ethoxybenzamide, 97%
Show More Show Less
EINECSNumber 213-346-4
RTECSNumber CV4900000
TSCA TSCA

RUO – Research Use Only