missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. E005825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E005825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E005825G Supplier TCI America Supplier No. E005825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 613-69-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00003316
InChI Key DUVJMSPTZMCSTQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2-ethoxy, o-ethoxybenzaldehyde, 2-ethoxy benzaldehyde, benzaldehyde, o-ethoxy, 2-ethoxy-benzaldehyde, benzaldehyde, ethoxy, ethoxy benzaldehyde, pubchem9726, ortho-ethoxybenzaldehyde, acmc-209ms1
PubChem CID 11950
IUPAC Name 2-ethoxybenzaldehyde
SMILES CCOC1=CC=CC=C1C=O

Specifications

Color Yellow
Boiling Point 249°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzaldehyde