missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzaldehyde, 97+%

Catalog No. AAA1503522
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1503522 100 g
AAA1503518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1503522 Supplier Thermo Scientific Chemicals Supplier No. A1503522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 613-69-4 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-69-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00003316
InChI Key DUVJMSPTZMCSTQ-UHFFFAOYSA-N
Synonym benzaldehyde, 2-ethoxy, o-ethoxybenzaldehyde, 2-ethoxy benzaldehyde, benzaldehyde, o-ethoxy, 2-ethoxy-benzaldehyde, benzaldehyde, ethoxy, ethoxy benzaldehyde, pubchem9726, ortho-ethoxybenzaldehyde, acmc-209ms1
PubChem CID 11950
IUPAC Name 2-ethoxybenzaldehyde
SMILES CCOC1=CC=CC=C1C=O

Specifications

Density 1.077
Boiling Point 249°C
Flash Point 91°C (195°F)
Refractive Index 1.543
Quantity 100 g
Beilstein 1937270
Sensitivity Air Sensitive
Formula Weight 150.18
Percent Purity ≥97%
Chemical Name or Material 2-Ethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-349-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only