Learn More
2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide
CAS: 14077-58-8 | C4H7ClO2 | 122.548 g/mol
Supplier: Thermo Scientific Chemicals H3444314
Description
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.
Solubility
Insoluble in water.
Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications
| 2-Ethoxyacetyl chloride | |
| 14077-58-8 | |
| CH3CH2OCH2COCl | |
| 25 g | |
| ethoxyacetyl chloride, ethoxyacetylchloride, ethoxy-acetyl chloride, acetyl chloride,2-ethoxy, 2-ethoxyacetylchloride, ethoxy acetyl chloride, ethoxyacetic acid chloride, 2-ethoxy-acetyl chloride, acetyl chloride, 2-ethoxy | |
| ZPMWWAIBJJFPPQ-UHFFFAOYSA-N | |
| 2-ethoxyacetyl chloride | |
| 84189 | |
| 97% |
| stabilized with 0.3% Magnesium oxide | |
| C4H7ClO2 | |
| MFCD11170566 | |
| UN2920 | |
| Insoluble in water. | |
| CCOCC(=O)Cl | |
| 122.548 | |
| 122.55 |
Chemical Identifiers
| 14077-58-8 | |
| 122.548 | |
| ZPMWWAIBJJFPPQ-UHFFFAOYSA-N | |
| 2-ethoxyacetyl chloride |
| C4H7ClO2 | |
| MFCD11170566 | |
| 84189 | |
| CCOCC(=O)Cl |
Safety and Handling
GHS H Statement
H314-H226-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 237-930-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only