missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide

Catalog No. AAH3444306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3444306 5 g
AAH3444314 25 g
AAH3444322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3444306 Supplier Thermo Scientific Chemicals Supplier No. H3444306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 14077-58-8 | C4H7ClO2 | 122.548 g/mol

It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

Solubility
Insoluble in water.

Notes
Reacts with strong oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 14077-58-8
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD11170566
InChI Key ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
PubChem CID 84189
IUPAC Name 2-ethoxyacetyl chloride
SMILES CCOCC(=O)Cl

Specifications

Linear Formula CH3CH2OCH2COCl
Quantity 5 g
UN Number UN2920
Synonym ethoxyacetyl chloride, ethoxyacetylchloride, ethoxy-acetyl chloride, acetyl chloride,2-ethoxy, 2-ethoxyacetylchloride, ethoxy acetyl chloride, ethoxyacetic acid chloride, 2-ethoxy-acetyl chloride, acetyl chloride, 2-ethoxy
Solubility Information Insoluble in water.
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material 2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide
Hazard Category H226-H314-H332-H335
Hazard Statement GHS H Statement
H314-H226-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 237-930-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only