missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$156.59 - $156.59
Chemical Identifiers
| CAS | 123291-97-4 |
|---|---|
| Molecular Formula | C9H13BO3 |
| Molecular Weight (g/mol) | 180.01 |
| MDL Number | MFCD00196832 |
| InChI Key | KBGGTPGSWHNQEX-UHFFFAOYSA-N |
| PubChem CID | 4615191 |
| IUPAC Name | (2-ethoxy-5-methylphenyl)boronic acid |
| SMILES | CCOC1=C(C=C(C)C=C1)B(O)O |
Chemical Identifiers
| 123291-97-4 | |
| 180.01 | |
| KBGGTPGSWHNQEX-UHFFFAOYSA-N | |
| (2-ethoxy-5-methylphenyl)boronic acid |
| C9H13BO3 | |
| MFCD00196832 | |
| 4615191 | |
| CCOC1=C(C=C(C)C=C1)B(O)O |
Specifications
| 123291-97-4 | |
| White-Yellow | |
| MFCD00196832 | |
| 2-ethoxy-5-methylbenzeneboronic acid, 2-ethoxy-5-methylphenyl boronic acid, 2-ethoxy-5-methyl-phenyl boronic acid, 2-ethoxy-5-methylphenylboronicacid, 4-ethoxytoluene-3-boronic acid, acmc-1c5pf, contains varying amounts of anhydride | |
| CCOC1=C(C=C(C)C=C1)B(O)O | |
| 180.01 | |
| 180.01 | |
| 2-Ethoxy-5-methylphenylboronic Acid (contains varying amounts of Anhydride) |
| 94°C | |
| C9H13BO3 | |
| 1 g | |
| KBGGTPGSWHNQEX-UHFFFAOYSA-N | |
| (2-ethoxy-5-methylphenyl)boronic acid | |
| 4615191 | |
| Crystalline Powder |
Safety and Handling
TSCA : No