missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-4-methylphenol, 95%

CAS: 2563-07-7 | C9H12O2 | 152.19 g/mol

Supplier:  Thermo Scientific Chemicals L0889809

 View more versions of this product

Catalog No. AAL0889809

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethoxy-4-methylphenol
28°C to 30°C
C5H2Cl2N2O3
MFCD00043697
1865448
LWOLKOLHQCMQDE-UHFFFAOYSA-N
2,6-dichloro-4-nitro-1-oxidopyridin-1-ium
2784721
95%
2563-07-7
103°C (3 mmHg)
1.523
10 g
2,6-dichloro-4-nitropyridine 1-oxide, 2,6-dichloro-4-nitropyridine n-oxide, 2,6-dichloro-4-nitropyridine-n-oxide, 2,6-dichloro-4-nitropyridine-1-oxide, 2,6-dichloro-4-nitropyridin-1-ium-1-olate, 2,6-dichloro-4-nitro-pyridine-n-oxide, pubchem5533, acmc-209usu, 2,6-dichloro-4-nitropyridin-1-ol, 2-chloro-4-nitro-pyridine-n-oxide
C1=C(C=C([N+](=C1Cl)[O-])Cl)[N+](=O)[O-]
208.98
152.19

Chemical Identifiers

2563-07-7
MFCD00043697
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only