missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecylhexadecan-1-ol 93.0+%, TCI America™

Catalog No. D532225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D532225G 25 g
D5322250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D532225G Supplier TCI America Supplier No. D532225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72388-18-2
Molecular Formula C28H58O
Molecular Weight (g/mol) 410.77
MDL Number MFCD27665210
InChI Key DEMBLPGWNXUBIQ-UHFFFAOYNA-N
Synonym 2-Dodecylhexadecyl Alcohol
PubChem CID 3018340
IUPAC Name 2-dodecylhexadecan-1-ol
SMILES CCCCCCCCCCCCCCC(CO)CCCCCCCCCCCC

Specifications

Melting Point 35°C
Color White
Quantity 25 g
Formula Weight 410.77
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Dodecylhexadecan-1-ol
EINECSNumber (2)-0217
TSCA Yes
Recommended Storage Refrigerator