missing translation for 'onlineSavingsMsg'
Learn More

2-Dodecen-1-ylsuccinic Anhydride (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. D097625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D097625G 25 g
D0976500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D097625G Supplier TCI America Supplier No. D097625G

Chemical Identifiers

CAS 19780-11-1
Molecular Formula C16H26O3
Molecular Weight (g/mol) 266.38
MDL Number MFCD00005528
InChI Key UYCICMIUKYEYEU-ZHACJKMWNA-N
PubChem CID 5362708
IUPAC Name 3-[(2E)-dodec-2-en-1-yl]oxolane-2,5-dione
SMILES CCCCCCCCC\C=C\CC1CC(=O)OC1=O

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystal-Lump at 20°C
UN Number 3077
Formula Weight 266.38
Percent Purity ≥95.0% (T)
Chemical Name or Material 2-Dodecen-1-ylsuccinic Anhydride (cis- and trans- mixture)
EINECSNumber (2)-0852
RTECSNumber WN1225000
TSCA Yes