missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. D43571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43571G 1 g
D43575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43571G Supplier TCI America Supplier No. D43571G

Chemical Identifiers

CAS 185990-03-8
Molecular Formula C14H23BO2Si
Molecular Weight (g/mol) 262.231
MDL Number MFCD05664111
InChI Key ARMSAQNLTKGMGM-UHFFFAOYSA-N
Synonym (Dimethylphenylsilyl)boronic Acid Pinacol Ester
PubChem CID 10199126
IUPAC Name dimethyl-phenyl-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane
SMILES B1(OC(C(O1)(C)C)(C)C)[Si](C)(C)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 262.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane