missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylcarbamoyl)benzeneboronic acid, 95%

Catalog No. p-7035060
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52765MD 250 mg
AAH5276503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52765MD Supplier Thermo Scientific Chemicals Supplier No. H52765MD
Only null left

CAS: 874219-16-6 | C9H12BNO3 | 193.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 874219-16-6
Molecular Formula C9H12BNO3
Molecular Weight (g/mol) 193.009
MDL Number MFCD03425959
InChI Key NZIOVLXULSCCSG-UHFFFAOYSA-N
Synonym 2-dimethylcarbamoyl phenyl boronic acid, 2-n,n-dimethylaminocarbonyl phenylboronic acid, 2-dimethylcarbamoyl phenylboronic acid, 2-dimethylaminocarbonyl benzeneboronic acid, boronic acid,b-2-dimethylamino carbonyl phenyl, 2-dimethylcarbamoyl benzeneboronic acid, acmc-209qkh, 2-n,n-dimethylaminocarbonyl benzeneboronic acid, 2-dimethylcarbamoyl phenyl boronicacid
PubChem CID 44119823
IUPAC Name [2-(dimethylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1C(=O)N(C)C)(O)O

Specifications

Melting Point 108°C to 110°C
Quantity 250 mg
Formula Weight 193.01
Percent Purity 95%
Chemical Name or Material 2-(Dimethylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only