missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminopyridine 98.0+%, TCI America™

Catalog No. p-7126873
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D168025G 25 g
D1680500G 500 g
2 options

Catalog No. D168025G

Supplier: TCI America D168025G

Only null left

Chemical Identifiers

CAS 5683-33-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006261
InChI Key PSHKMPUSSFXUIA-UHFFFAOYSA-N
Synonym 2-dimethylaminopyridine, 2-dimethylaminopyridin, dimethylaminopyridine, 2-dimethylamino pyridine, 2-dimethylaminopyridine cobalt complex, 2-pyridinamine, n,n-dimethyl, dimethylamino-2 pyridine, dimethylaminopyridin, unii-18q95u3z8h, pyridine, 2-dimethylamino
PubChem CID 21885
IUPAC Name N,N-dimethylpyridin-2-amine
SMILES CN(C)C1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminopyridine
RTECSNumber US9200000
TSCA No